missing translation for 'onlineSavingsMsg'
Learn More

2,5-Thiophenedicarboxylic Acid 98.0+%, TCI America™

Catalog No. p-7137170
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T23475G 5 g
T234725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T23475G Supplier TCI America Supplier No. T23475G
Only null left

Chemical Identifiers

CAS 4282-31-9
Molecular Formula C6H4O4S
Molecular Weight (g/mol) 172.154
MDL Number MFCD00016896
InChI Key YCGAZNXXGKTASZ-UHFFFAOYSA-N
Synonym 2,5-thiophenedicarboxylic acid, 2,5-dicarboxythiophene, unii-5q8a2g9y8a, 2,5-thiophene dicarboxylic acid, pubchem5580, maybridge4_002021, acmc-1aj5p, ksc182i5d, 2,5-thiophene-dicarboxylic acid, thiophene-2.5-dicarboxylic acid
PubChem CID 20273
IUPAC Name thiophene-2,5-dicarboxylic acid
SMILES C1=C(SC(=C1)C(=O)O)C(=O)O

Specifications

Melting Point 359°C
Color White-Yellow
Quantity 5 g
Formula Weight 172.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Thiophenedicarboxylic Acid