missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylbenzoic Acid 98.0+%, TCI America™

Catalog No. p-7126726
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D143310G 10 g
D143325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D143310G Supplier TCI America Supplier No. D143310G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 610-72-0 | C9H10O2 | 150.177 g/mol

Chemical Identifiers

CAS 610-72-0
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00002482
InChI Key XZRHNAFEYMSXRG-UHFFFAOYSA-N
Synonym benzoic acid, 2,5-dimethyl, isoxylic acid, 2,5-dimethyl benzoic acid, unii-rke5pmk0h6, 2,5-dimethyl-benzoic acid, 2-carboxy-1,4-dimethylbenzene, rke5pmk0h6, p-xylene-2-carboxylic acid, 2,5-dimethylbenzenecarboxylic acid, 2,5-dimethylbenzoicacid
PubChem CID 11892
ChEBI CHEBI:64825
IUPAC Name 2,5-dimethylbenzoic acid
SMILES CC1=CC(=C(C=C1)C)C(=O)O

Specifications

Melting Point 134°C
Color White
Quantity 10 g
Physical Form Crystal-Powder at 20°C
Formula Weight 150.18
Percent Purity ≥98.0% (GC,T)
Grade Not Available
Chemical Name or Material 2,5-Dimethylbenzoic Acid