accessibility menu, dialog, popup

UserName

CAS RN 610-72-0

IUPAC Name: 2,5-dimethylbenzoic acid
Synonyms: benzoic acid, 2,5-dimethyl-
Molecular Weight (g/mol): 150.18
Molecular Formula: C9H10O2
InChi Key: XZRHNAFEYMSXRG-UHFFFAOYSA-N
SMILES: CC1=CC=C(C)C(=C1)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)