missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dimethylbenzoic Acid 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 134°C |
| Color | White |
| Quantity | 10 g |
| Physical Form | Crystal-Powder at 20°C |
| Formula Weight | 150.18 |
| Percent Purity | ≥98.0% (GC,T) |
| Grade | Not Available |
| Chemical Name or Material | 2,5-Dimethylbenzoic Acid |
Safety and Handling
| TSCA | Yes |