missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2',5'-Dimethylacetophenone 98.0+%, TCI America™
Supplier: TCI America D1880250ML
Specifications
| 2′,5′-Dimethylacetophenone | |
| Yellow | |
| C10H12O | |
| 250 mL | |
| AWKBVLVKQQRRFQ-UHFFFAOYSA-N | |
| 1-(2,5-dimethylphenyl)ethanone | |
| 75061 | |
| ≥98.0% (GC) |
| 2142-73-6 | |
| 112°C | |
| MFCD00014988 | |
| 2',5'-dimethylacetophenone, 2,5-dimethylacetophenone, 1-2,5-dimethylphenyl ethanone, 1-2,5-dimethylphenyl ethan-1-one, ethanone, 1-2,5-dimethylphenyl, acetophenone, 2',5'-dimethyl, 1-acetyl-2,5-dimethylbenzene, 2-acetyl-p-xylene, acmc-1cliy, 2',5'dimethylacetophenone | |
| CC1=CC(=C(C=C1)C)C(=O)C | |
| 148.205 | |
| 148.21 | |
| Liquid |
Chemical Identifiers
| 2142-73-6 | |
| 148.205 | |
| AWKBVLVKQQRRFQ-UHFFFAOYSA-N | |
| 75061 | |
| CC1=CC(=C(C=C1)C)C(=O)C |
| C10H12O | |
| MFCD00014988 | |
| 2',5'-dimethylacetophenone, 2,5-dimethylacetophenone, 1-2,5-dimethylphenyl ethanone, 1-2,5-dimethylphenyl ethan-1-one, ethanone, 1-2,5-dimethylphenyl, acetophenone, 2',5'-dimethyl, 1-acetyl-2,5-dimethylbenzene, 2-acetyl-p-xylene, acmc-1cliy, 2',5'dimethylacetophenone | |
| 1-(2,5-dimethylphenyl)ethanone |
Safety and Handling
TSCA : Yes