missing translation for 'onlineSavingsMsg'
Learn More

2',5'-Dimethylacetophenone 98.0+%, TCI America™

Catalog No. p-7126989
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D188025ML 25 mL
D1880250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D188025ML Supplier TCI America Supplier No. D188025ML

Chemical Identifiers

CAS 2142-73-6
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.205
MDL Number MFCD00014988
InChI Key AWKBVLVKQQRRFQ-UHFFFAOYSA-N
Synonym 2',5'-dimethylacetophenone, 2,5-dimethylacetophenone, 1-2,5-dimethylphenyl ethanone, 1-2,5-dimethylphenyl ethan-1-one, ethanone, 1-2,5-dimethylphenyl, acetophenone, 2',5'-dimethyl, 1-acetyl-2,5-dimethylbenzene, 2-acetyl-p-xylene, acmc-1cliy, 2',5'dimethylacetophenone
PubChem CID 75061
IUPAC Name 1-(2,5-dimethylphenyl)ethanone
SMILES CC1=CC(=C(C=C1)C)C(=O)C

Specifications

Color Yellow
Boiling Point 112°C
Quantity 25 mL
Formula Weight 148.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2′,5′-Dimethylacetophenone