Learn More
2,5-Dimethoxyphenylacetic acid, 99%
CAS: 1758-25-4 | C10H12O4 | 196.202 g/mol
$118.11 - $377.78
Chemical Identifiers
| CAS | 1758-25-4 |
|---|---|
| Molecular Formula | C10H12O4 |
| Molecular Weight (g/mol) | 196.202 |
| MDL Number | MFCD00004322 |
| InChI Key | BBZDYQUXRFATHZ-UHFFFAOYSA-N |
| Synonym | 2,5-dimethoxyphenylacetic acid, 2-2,5-dimethoxyphenyl acetic acid, 2,5-dimethoxyphenyl acetic acid, benzeneacetic acid, 2,5-dimethoxy, acetic acid, 2,5-dimethoxyphenyl, 2,5-dimethoxyphenyl-acetic acid, acmc-209eai, 2,5-dimethoxyphenyl, rarechem al bo 0334, 2,5-dimethoxy-phenethyl acid |
| PubChem CID | 74469 |
| IUPAC Name | 2-(2,5-dimethoxyphenyl)acetic acid |
| SMILES | COC1=CC(=C(C=C1)OC)CC(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH2732806
|
Thermo Scientific Chemicals
H2732806 |
5 g |
Each for $118.11
|
|
|||||
|
AAH2732814
|
Thermo Scientific Chemicals
H2732814 |
25 g |
Each for $377.78
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1758-25-4 | |
| 196.202 | |
| BBZDYQUXRFATHZ-UHFFFAOYSA-N | |
| 74469 | |
| COC1=CC(=C(C=C1)OC)CC(=O)O |
| C10H12O4 | |
| MFCD00004322 | |
| 2,5-dimethoxyphenylacetic acid, 2-2,5-dimethoxyphenyl acetic acid, 2,5-dimethoxyphenyl acetic acid, benzeneacetic acid, 2,5-dimethoxy, acetic acid, 2,5-dimethoxyphenyl, 2,5-dimethoxyphenyl-acetic acid, acmc-209eai, 2,5-dimethoxyphenyl, rarechem al bo 0334, 2,5-dimethoxy-phenethyl acid | |
| 2-(2,5-dimethoxyphenyl)acetic acid |
Specifications
| 1758-25-4 | |
| C10H12O4 | |
| 5 g | |
| BBZDYQUXRFATHZ-UHFFFAOYSA-N | |
| 2-(2,5-dimethoxyphenyl)acetic acid | |
| 74469 | |
| 99% |
| 123°C to 125°C | |
| MFCD00004322 | |
| 2,5-dimethoxyphenylacetic acid, 2-2,5-dimethoxyphenyl acetic acid, 2,5-dimethoxyphenyl acetic acid, benzeneacetic acid, 2,5-dimethoxy, acetic acid, 2,5-dimethoxyphenyl, 2,5-dimethoxyphenyl-acetic acid, acmc-209eai, 2,5-dimethoxyphenyl, rarechem al bo 0334, 2,5-dimethoxy-phenethyl acid | |
| COC1=CC(=C(C=C1)OC)CC(=O)O | |
| 196.202 | |
| 196.2 | |
| 2,5-Dimethoxyphenylacetic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 217-151-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only