missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethoxyphenylacetic acid, 99%

Catalog No. AAH2732806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2732806 5 g
AAH2732814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2732806 Supplier Thermo Scientific Chemicals Supplier No. H2732806

CAS: 1758-25-4 | C10H12O4 | 196.202 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1758-25-4
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.202
MDL Number MFCD00004322
InChI Key BBZDYQUXRFATHZ-UHFFFAOYSA-N
Synonym 2,5-dimethoxyphenylacetic acid, 2-2,5-dimethoxyphenyl acetic acid, 2,5-dimethoxyphenyl acetic acid, benzeneacetic acid, 2,5-dimethoxy, acetic acid, 2,5-dimethoxyphenyl, 2,5-dimethoxyphenyl-acetic acid, acmc-209eai, 2,5-dimethoxyphenyl, rarechem al bo 0334, 2,5-dimethoxy-phenethyl acid
PubChem CID 74469
IUPAC Name 2-(2,5-dimethoxyphenyl)acetic acid
SMILES COC1=CC(=C(C=C1)OC)CC(=O)O

Specifications

Melting Point 123°C to 125°C
Quantity 5 g
Formula Weight 196.2
Percent Purity 99%
Chemical Name or Material 2,5-Dimethoxyphenylacetic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 217-151-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only