missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethoxybenzoic acid, 98+%

Catalog No. p-7020819
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1290106 5 g
AAA1290114 25 g
AAA1290122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1290106 Supplier Thermo Scientific Chemicals Supplier No. A1290106
Only null left

CAS: 2785-98-0 | C9H10O4 | 182.175 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2785-98-0
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.175
MDL Number MFCD00002436
InChI Key NYJBTJMNTNCTCP-UHFFFAOYSA-N
Synonym benzoic acid, 2,5-dimethoxy, 6-methoxy-m-anisic acid, 2,5-dimethoxy-benzoic acid, 2,5-dimethoxybenzoicacid, pubchem14413, acmc-1cit0, 2,5-dimethoxybenzoic acid
PubChem CID 76027
IUPAC Name 2,5-dimethoxybenzoic acid
SMILES COC1=CC(=C(C=C1)OC)C(=O)O

Specifications

Melting Point 75°C to 79°C
Odor Odorless
Quantity 5 g
Beilstein 2097993
Formula Weight 182.18
Percent Purity ≥98%
Chemical Name or Material 2,5-Dimethoxybenzoic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 220-503-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only