missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dihydroxyterephthalic Acid 98.0+%, TCI America™

Supplier:  TCI America D389925G

 View more versions of this product

Catalog No. D389925G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2,5-Dihydroxyterephthalic Acid
339°C
MFCD00132933
1,4-benzenedicarboxylic acid, 2,5-dihydroxy, 2,5-dihydroxyterephthalicacid, 2,5-dihydroxy-1,4-benzenedicarboxylic acid, 2,5-dihydroxybenzene-1,4-dicarboxylic acid, zlchem 699, acmc-1ayqi, 1, 2,5-dihydroxy, 2,5-dihydroxytelephthalic acid, #, 2,5-dihydroxyterephthalic acid
C1=C(C(=CC(=C1O)C(=O)O)O)C(=O)O
198.13
198.13
Crystalline Powder
610-92-4
C8H6O6
25 g
OYFRNYNHAZOYNF-UHFFFAOYSA-N
2,5-dihydroxyterephthalic acid
69131
≥98.0% (HPLC,T)

Chemical Identifiers

610-92-4
198.13
OYFRNYNHAZOYNF-UHFFFAOYSA-N
69131
C1=C(C(=CC(=C1O)C(=O)O)O)C(=O)O
C8H6O6
MFCD00132933
1,4-benzenedicarboxylic acid, 2,5-dihydroxy, 2,5-dihydroxyterephthalicacid, 2,5-dihydroxy-1,4-benzenedicarboxylic acid, 2,5-dihydroxybenzene-1,4-dicarboxylic acid, zlchem 699, acmc-1ayqi, 1, 2,5-dihydroxy, 2,5-dihydroxytelephthalic acid, #, 2,5-dihydroxyterephthalic acid
2,5-dihydroxyterephthalic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions