missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dihydroxyterephthalic Acid 98.0+%, TCI America™

Catalog No. p-7127983
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38995G 5 g
D389925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D38995G Supplier TCI America Supplier No. D38995G

Chemical Identifiers

CAS 610-92-4
Molecular Formula C8H6O6
Molecular Weight (g/mol) 198.13
MDL Number MFCD00132933
InChI Key OYFRNYNHAZOYNF-UHFFFAOYSA-N
Synonym 1,4-benzenedicarboxylic acid, 2,5-dihydroxy, 2,5-dihydroxyterephthalicacid, 2,5-dihydroxy-1,4-benzenedicarboxylic acid, 2,5-dihydroxybenzene-1,4-dicarboxylic acid, zlchem 699, acmc-1ayqi, 1, 2,5-dihydroxy, 2,5-dihydroxytelephthalic acid, #, 2,5-dihydroxyterephthalic acid
PubChem CID 69131
IUPAC Name 2,5-dihydroxyterephthalic acid
SMILES C1=C(C(=CC(=C1O)C(=O)O)O)C(=O)O

Specifications

Melting Point 339°C
Quantity 5 g
Formula Weight 198.13
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dihydroxyterephthalic Acid