missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dihydroxybenzoic Acid Sodium Salt Hydrate 98.0+%, TCI America™
Supplier: TCI America D489325G
Specifications
| 2,5-Dihydroxybenzoic Acid Sodium Salt Hydrate | |
| White-Yellow | |
| MFCD00150273 | |
| Gentisic Acid Sodium Salt | |
| C1=CC(=C(C=C1O)C(=O)[O-])O.[Na+] | |
| 176.103 | |
| 176.10 | |
| Crystalline Powder |
| 4955-90-2 | |
| C7H5NaO4 | |
| 25 g | |
| MOIJZWWOFOQFMH-UHFFFAOYSA-M | |
| sodium;2,5-dihydroxybenzoate | |
| 23663629 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 4955-90-2 | |
| 176.103 | |
| MOIJZWWOFOQFMH-UHFFFAOYSA-M | |
| 23663629 | |
| C1=CC(=C(C=C1O)C(=O)[O-])O.[Na+] |
| C7H5NaO4 | |
| MFCD00150273 | |
| Gentisic Acid Sodium Salt | |
| sodium;2,5-dihydroxybenzoate |
Safety and Handling
EINECSNumber : (3)-3067
RTECSNumber : LY3860000
TSCA : Yes