accessibility menu, dialog, popup

UserName

NaOHOHOO

CAS RN 4955-90-2

IUPAC Name: sodium 2,5-dihydroxybenzoate
Synonyms: sodium gentisate 龙胆酸钠
Molecular Weight (g/mol): 176.1
Molecular Formula: C7H5NaO4
InChi Key: MOIJZWWOFOQFMH-UHFFFAOYSA-M
SMILES: [Na+].OC1=CC=C(O)C(=C1)C([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results