missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dihydroxybenzoic Acid Sodium Salt Hydrate 98.0+%, TCI America™

Catalog No. D48935G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48935G 5 g
D489325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D48935G Supplier TCI America Supplier No. D48935G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4955-90-2
Molecular Formula C7H5NaO4
Molecular Weight (g/mol) 176.103
MDL Number MFCD00150273
InChI Key MOIJZWWOFOQFMH-UHFFFAOYSA-M
Synonym Gentisic Acid Sodium Salt
PubChem CID 23663629
IUPAC Name sodium;2,5-dihydroxybenzoate
SMILES C1=CC(=C(C=C1O)C(=O)[O-])O.[Na+]

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 176.10
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dihydroxybenzoic Acid Sodium Salt Hydrate
EINECSNumber (3)-3067
RTECSNumber LY3860000
TSCA Yes