missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluorophenyl isocyanate, 98%

Catalog No. p-7046716
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL15996MD 250 mg
AAL1599603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL15996MD Supplier Thermo Scientific Chemicals Supplier No. L15996MD
Only null left

CAS: 39718-32-6 | C7H3F2NO | 155.104 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 39718-32-6
Molecular Formula C7H3F2NO
Molecular Weight (g/mol) 155.104
MDL Number MFCD00013843
InChI Key SNHIIFOXCRYGGY-UHFFFAOYSA-N
PubChem CID 2733266
IUPAC Name 1,4-difluoro-2-isocyanatobenzene
SMILES C1=CC(=C(C=C1F)N=C=O)F

Specifications

Density 1.323
Boiling Point 78°C (33 mmHg)
Flash Point 56°C (132°F)
Refractive Index 1.494
Quantity 250 mg
UN Number UN3080
Beilstein 2091753
Sensitivity Moisture sensitive
Synonym 2,5-difluorophenyl isocyanate, 2,5-difluorophenylisocyanate, benzene, 1,4-difluoro-2-isocyanato, isocyanic acid 2,5-difluorophenyl ester, 2,5-difluorobenzenisocyanate, pubchem5022, acmc-1ahno, 2,5-difluoro-phenyl-isocyanate, benzene,1,4-difluoro-2-isocyanato
Formula Weight 155.11
Percent Purity 98%
Chemical Name or Material 2,5-Difluorophenyl isocyanate
Show More Show Less
Hazard Category H226-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H300-H330-H334-H226-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ISOCYANATES, TOXIC, FLAMMABLE, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only