missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Difluorobenzylamine 98.0+%, TCI America™
$552.52 - $552.52
Chemical Identifiers
| CAS | 85118-06-5 |
|---|---|
| Molecular Formula | C7H7F2N |
| Molecular Weight (g/mol) | 143.14 |
| MDL Number | MFCD00010143 |
| InChI Key | GDFBHCMFIUBEQT-UHFFFAOYSA-N |
| Synonym | 2,5-difluorobenzylamine, 2,5-difluorophenyl methanamine, benzenemethanamine, 2,5-difluoro, 2,5-difluorobenzyl amine, 1-2,5-difluorophenyl methanamine, 2, 5-difluorobenzylamine, 2,5-difluorophenyl methylamine, pubchem4401, 2,5-difluoro-benzylamine, acmc-209q4p |
| PubChem CID | 123589 |
| IUPAC Name | 1-(2,5-difluorophenyl)methanamine |
| SMILES | NCC1=CC(F)=CC=C1F |
Chemical Identifiers
| 85118-06-5 | |
| 143.14 | |
| GDFBHCMFIUBEQT-UHFFFAOYSA-N | |
| 123589 | |
| NCC1=CC(F)=CC=C1F |
| C7H7F2N | |
| MFCD00010143 | |
| 2,5-difluorobenzylamine, 2,5-difluorophenyl methanamine, benzenemethanamine, 2,5-difluoro, 2,5-difluorobenzyl amine, 1-2,5-difluorophenyl methanamine, 2, 5-difluorobenzylamine, 2,5-difluorophenyl methylamine, pubchem4401, 2,5-difluoro-benzylamine, acmc-209q4p | |
| 1-(2,5-difluorophenyl)methanamine |
Specifications
| 85118-06-5 | |
| 78°C | |
| MFCD00010143 | |
| 2735 | |
| GDFBHCMFIUBEQT-UHFFFAOYSA-N | |
| 1-(2,5-difluorophenyl)methanamine | |
| 123589 | |
| ≥98.0% (GC,T) | |
| 2,5-Difluorobenzylamine |
| Yellow | |
| C7H7F2N | |
| 5 g | |
| 2,5-difluorobenzylamine, 2,5-difluorophenyl methanamine, benzenemethanamine, 2,5-difluoro, 2,5-difluorobenzyl amine, 1-2,5-difluorophenyl methanamine, 2, 5-difluorobenzylamine, 2,5-difluorophenyl methylamine, pubchem4401, 2,5-difluoro-benzylamine, acmc-209q4p | |
| NCC1=CC(F)=CC=C1F | |
| 143.14 | |
| 143.14 | |
| Liquid |
Safety and Handling
TSCA : No