missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluorobenzylamine, 97%

Catalog No. AAA1653003
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1653003 1 g
AAA1653006 5 g
AAA1653014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1653003 Supplier Thermo Scientific Chemicals Supplier No. A1653003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 85118-06-5 | C7H7F2N | 143.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 85118-06-5
Molecular Formula C7H7F2N
Molecular Weight (g/mol) 143.14
MDL Number MFCD00010143
InChI Key GDFBHCMFIUBEQT-UHFFFAOYSA-N
Synonym 2,5-difluorobenzylamine, 2,5-difluorophenyl methanamine, benzenemethanamine, 2,5-difluoro, 2,5-difluorobenzyl amine, 1-2,5-difluorophenyl methanamine, 2, 5-difluorobenzylamine, 2,5-difluorophenyl methylamine, pubchem4401, 2,5-difluoro-benzylamine, acmc-209q4p
PubChem CID 123589
IUPAC Name (2,5-difluorophenyl)methanamine
SMILES NCC1=CC(F)=CC=C1F

Specifications

Density 1.222
Boiling Point 76°C to 78°C (15 mmHg)
Flash Point 75°C (167°F)
Refractive Index 1.495
Quantity 1 g
UN Number UN2735
Beilstein 3539280
Sensitivity Air Sensitive
Formula Weight 143.14
Percent Purity 97%
Chemical Name or Material 2,5-Difluorobenzylamine
Show More Show Less
Hazard Category H227-H314
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 285-658-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only