missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Difluorobenzyl Bromide 98.0+%, TCI America™
$61.51 - $188.27
Chemical Identifiers
| CAS | 85117-99-3 |
|---|---|
| Molecular Formula | C7H5BrF2 |
| Molecular Weight (g/mol) | 207.02 |
| MDL Number | MFCD00009897 |
| InChI Key | ONWGSWNHQZYCFK-UHFFFAOYSA-N |
| Synonym | 2,5-difluorobenzyl bromide, 2-bromomethyl-1,4-difluorobenzene, 2,5-difluorobenzylbromide, alpha-bromo-2,5-difluorotoluene, benzene, 2-bromomethyl-1,4-difluoro, timtec-bb sbb006680, 2-bromomethyl-1,4-difluoro-benzene, pubchem10213, 2,2-difluorobenzylbromide, acmc-209q4i |
| PubChem CID | 522830 |
| IUPAC Name | 2-(bromomethyl)-1,4-difluorobenzene |
| SMILES | FC1=CC=C(F)C(CBr)=C1 |
Chemical Identifiers
| 85117-99-3 | |
| 207.02 | |
| ONWGSWNHQZYCFK-UHFFFAOYSA-N | |
| 522830 | |
| FC1=CC=C(F)C(CBr)=C1 |
| C7H5BrF2 | |
| MFCD00009897 | |
| 2,5-difluorobenzyl bromide, 2-bromomethyl-1,4-difluorobenzene, 2,5-difluorobenzylbromide, alpha-bromo-2,5-difluorotoluene, benzene, 2-bromomethyl-1,4-difluoro, timtec-bb sbb006680, 2-bromomethyl-1,4-difluoro-benzene, pubchem10213, 2,2-difluorobenzylbromide, acmc-209q4i | |
| 2-(bromomethyl)-1,4-difluorobenzene |
Specifications
| 85117-99-3 | |
| C7H5BrF2 | |
| 5 g | |
| 2,5-difluorobenzyl bromide, 2-bromomethyl-1,4-difluorobenzene, 2,5-difluorobenzylbromide, alpha-bromo-2,5-difluorotoluene, benzene, 2-bromomethyl-1,4-difluoro, timtec-bb sbb006680, 2-bromomethyl-1,4-difluoro-benzene, pubchem10213, 2,2-difluorobenzylbromide, acmc-209q4i | |
| FC1=CC=C(F)C(CBr)=C1 | |
| 207.02 | |
| 207.02 | |
| Liquid |
| Yellow | |
| MFCD00009897 | |
| 2924 | |
| ONWGSWNHQZYCFK-UHFFFAOYSA-N | |
| 2-(bromomethyl)-1,4-difluorobenzene | |
| 522830 | |
| ≥98.0% (GC) | |
| 2,5-Difluorobenzyl Bromide |
Safety and Handling
TSCA : No