missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Difluorobenzoyl Chloride 98.0+%, TCI America™
$61.51 - $206.90
Chemical Identifiers
| CAS | 35730-09-7 |
|---|---|
| Molecular Formula | C7H3ClF2O |
| Molecular Weight (g/mol) | 176.547 |
| MDL Number | MFCD00009929 |
| InChI Key | RLRUKKDFNWXXRT-UHFFFAOYSA-N |
| PubChem CID | 588082 |
| IUPAC Name | 2,5-difluorobenzoyl chloride |
| SMILES | C1=CC(=C(C=C1F)C(=O)Cl)F |
Chemical Identifiers
| 35730-09-7 | |
| 176.547 | |
| RLRUKKDFNWXXRT-UHFFFAOYSA-N | |
| 2,5-difluorobenzoyl chloride |
| C7H3ClF2O | |
| MFCD00009929 | |
| 588082 | |
| C1=CC(=C(C=C1F)C(=O)Cl)F |
Specifications
| 35730-09-7 | |
| 93°C | |
| MFCD00009929 | |
| 2920 | |
| RLRUKKDFNWXXRT-UHFFFAOYSA-N | |
| 2,5-difluorobenzoyl chloride | |
| 588082 | |
| ≥98.0% (GC,T) | |
| 2,5-Difluorobenzoyl Chloride |
| Yellow | |
| C7H3ClF2O | |
| 5 g | |
| 2,5-difluorobenzoylchloride, benzoyl chloride, 2,5-difluoro, benzoyl chloride,2,5-difluoro, acmc-1afez, 2,5-difluoro benzoyl chloride, 2,5-difluoro-benzoyl chloride, 2,5-difluorobenzoyl chloride | |
| C1=CC(=C(C=C1F)C(=O)Cl)F | |
| 176.547 | |
| 176.55 | |
| Liquid |
Safety and Handling
TSCA : No