missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Difluorobenzoyl Chloride 98.0+%, TCI America™
Supplier: TCI America D340625G
Specifications
| 2,5-Difluorobenzoyl Chloride | |
| Yellow | |
| C7H3ClF2O | |
| 25 g | |
| 2,5-difluorobenzoylchloride, benzoyl chloride, 2,5-difluoro, benzoyl chloride,2,5-difluoro, acmc-1afez, 2,5-difluoro benzoyl chloride, 2,5-difluoro-benzoyl chloride, 2,5-difluorobenzoyl chloride | |
| C1=CC(=C(C=C1F)C(=O)Cl)F | |
| 176.547 | |
| 176.55 | |
| Liquid |
| 35730-09-7 | |
| 93°C | |
| MFCD00009929 | |
| 2920 | |
| RLRUKKDFNWXXRT-UHFFFAOYSA-N | |
| 2,5-difluorobenzoyl chloride | |
| 588082 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 35730-09-7 | |
| 176.547 | |
| RLRUKKDFNWXXRT-UHFFFAOYSA-N | |
| 2,5-difluorobenzoyl chloride |
| C7H3ClF2O | |
| MFCD00009929 | |
| 588082 | |
| C1=CC(=C(C=C1F)C(=O)Cl)F |
Safety and Handling
TSCA : No