missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluorobenzotrifluoride 98.0+%, TCI America™

Catalog No. D49331G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49331G 1 g
D49335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D49331G Supplier TCI America Supplier No. D49331G

Chemical Identifiers

CAS 393-38-4
Molecular Formula C7H3F5
Molecular Weight (g/mol) 182.09
MDL Number MFCD00042494
InChI Key BLDOSVXRDMTZQE-UHFFFAOYSA-N
Synonym 2,5-difluorobenzotrifluoride, 1,4-difluoro-2-trifluoromethyl benzene, 1,4-difluoro-2-trifluoromethyl-benzene, benzene, 1,4-difluoro-2-trifluoromethyl, pubchem1650, 1-trifluoromethyl-2,5-difluorobenzene
PubChem CID 11111097
IUPAC Name 1,4-difluoro-2-(trifluoromethyl)benzene
SMILES FC1=CC(=C(F)C=C1)C(F)(F)F

Specifications

Melting Point -14°C
Color Yellow
Boiling Point 108°C
Quantity 1 g
UN Number 1993
Formula Weight 182.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Difluorobenzotrifluoride