accessibility menu, dialog, popup

UserName

CAS RN 393-38-4

IUPAC Name: 1,4-difluoro-2-(trifluoromethyl)benzene
Synonyms: 1,4-difluoro-2-(trifluoromethyl)benzene
Molecular Weight (g/mol): 182.09
Molecular Formula: C7H3F5
InChi Key: BLDOSVXRDMTZQE-UHFFFAOYSA-N
SMILES: FC1=CC(=C(F)C=C1)C(F)(F)F
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)