missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Difluorobenzenesulfonamide 98.0+%, TCI America™
$134.21 - $451.10
Chemical Identifiers
| CAS | 120022-63-1 |
|---|---|
| Molecular Formula | C6H5F2NO2S |
| Molecular Weight (g/mol) | 193.168 |
| MDL Number | MFCD00798115 |
| InChI Key | OLMFEUWXDKZGOO-UHFFFAOYSA-N |
| Synonym | 2,5-difluorobenzenesulphonamide, 2,5-difluorobenzene-1-sulfonamide, benzenesulfonamide, 2,5-difluoro, 2,5-dfbs, acmc-20ai8p, 2,5-difluoro-benzenesulfonamide, benzenesulfonamide,2,5-difluoro, 2,5-bis fluoranyl benzenesulfonamide, 2,5-difluorobenzenesulfonamide |
| PubChem CID | 147822 |
| IUPAC Name | 2,5-difluorobenzenesulfonamide |
| SMILES | C1=CC(=C(C=C1F)S(=O)(=O)N)F |
Chemical Identifiers
| 120022-63-1 | |
| 193.168 | |
| OLMFEUWXDKZGOO-UHFFFAOYSA-N | |
| 147822 | |
| C1=CC(=C(C=C1F)S(=O)(=O)N)F |
| C6H5F2NO2S | |
| MFCD00798115 | |
| 2,5-difluorobenzenesulphonamide, 2,5-difluorobenzene-1-sulfonamide, benzenesulfonamide, 2,5-difluoro, 2,5-dfbs, acmc-20ai8p, 2,5-difluoro-benzenesulfonamide, benzenesulfonamide,2,5-difluoro, 2,5-bis fluoranyl benzenesulfonamide, 2,5-difluorobenzenesulfonamide | |
| 2,5-difluorobenzenesulfonamide |
Specifications
| 120022-63-1 | |
| White | |
| MFCD00798115 | |
| 2,5-difluorobenzenesulphonamide, 2,5-difluorobenzene-1-sulfonamide, benzenesulfonamide, 2,5-difluoro, 2,5-dfbs, acmc-20ai8p, 2,5-difluoro-benzenesulfonamide, benzenesulfonamide,2,5-difluoro, 2,5-bis fluoranyl benzenesulfonamide, 2,5-difluorobenzenesulfonamide | |
| C1=CC(=C(C=C1F)S(=O)(=O)N)F | |
| 193.168 | |
| 193.17 | |
| Crystalline Powder |
| 137°C | |
| C6H5F2NO2S | |
| 1 g | |
| OLMFEUWXDKZGOO-UHFFFAOYSA-N | |
| 2,5-difluorobenzenesulfonamide | |
| 147822 | |
| ≥98.0% (GC,N) | |
| 2,5-Difluorobenzenesulfonamide |
Safety and Handling
TSCA : No