accessibility menu, dialog, popup

UserName

NH2SOOFF

CAS RN 120022-63-1

IUPAC Name: 2,5-difluorobenzene-1-sulfonamide
Synonyms: 2,5-difluorobenzenesulfonamide
Molecular Weight (g/mol): 193.17
Molecular Formula: C6H5F2NO2S
InChi Key: OLMFEUWXDKZGOO-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C1=CC(F)=CC=C1F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results