missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluorobenzenesulfonamide 98.0+%, TCI America™

Catalog No. D40605G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40605G 5 g
D40601G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D40605G Supplier TCI America Supplier No. D40605G
Only null left

Chemical Identifiers

CAS 120022-63-1
Molecular Formula C6H5F2NO2S
Molecular Weight (g/mol) 193.168
MDL Number MFCD00798115
InChI Key OLMFEUWXDKZGOO-UHFFFAOYSA-N
Synonym 2,5-difluorobenzenesulphonamide, 2,5-difluorobenzene-1-sulfonamide, benzenesulfonamide, 2,5-difluoro, 2,5-dfbs, acmc-20ai8p, 2,5-difluoro-benzenesulfonamide, benzenesulfonamide,2,5-difluoro, 2,5-bis fluoranyl benzenesulfonamide, 2,5-difluorobenzenesulfonamide
PubChem CID 147822
IUPAC Name 2,5-difluorobenzenesulfonamide
SMILES C1=CC(=C(C=C1F)S(=O)(=O)N)F

Specifications

Melting Point 137°C
Color White
Quantity 5 g
Formula Weight 193.17
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Difluorobenzenesulfonamide