missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Difluoroaniline 98.0+%, TCI America™
$75.08 - $240.64
Chemical Identifiers
| CAS | 367-30-6 |
|---|---|
| Molecular Formula | C6H5F2N |
| Molecular Weight (g/mol) | 129.11 |
| MDL Number | MFCD00007651 |
| InChI Key | YNOOQIUSYGWMSS-UHFFFAOYSA-N |
| Synonym | benzenamine, 2,5-difluoro, 2,5-difluorobenzenamine, 2,5-difluoro-phenylamine, aniline, 2,5-difluoro, 2.5-difluoroaniline, aniline,5-difluoro, pubchem1531, 2,5-difluoro-aniline, 2,5-difluorophenylamine, benzenamine,5-difluoro |
| PubChem CID | 67775 |
| IUPAC Name | 2,5-difluoroaniline |
| SMILES | C1=CC(=C(C=C1F)N)F |
Chemical Identifiers
| 367-30-6 | |
| 129.11 | |
| YNOOQIUSYGWMSS-UHFFFAOYSA-N | |
| 67775 | |
| C1=CC(=C(C=C1F)N)F |
| C6H5F2N | |
| MFCD00007651 | |
| benzenamine, 2,5-difluoro, 2,5-difluorobenzenamine, 2,5-difluoro-phenylamine, aniline, 2,5-difluoro, 2.5-difluoroaniline, aniline,5-difluoro, pubchem1531, 2,5-difluoro-aniline, 2,5-difluorophenylamine, benzenamine,5-difluoro | |
| 2,5-difluoroaniline |
Specifications
| 367-30-6 | |
| C6H5F2N | |
| 5 g | |
| benzenamine, 2,5-difluoro, 2,5-difluorobenzenamine, 2,5-difluoro-phenylamine, aniline, 2,5-difluoro, 2.5-difluoroaniline, aniline,5-difluoro, pubchem1531, 2,5-difluoro-aniline, 2,5-difluorophenylamine, benzenamine,5-difluoro | |
| C1=CC(=C(C=C1F)N)F | |
| 129.11 | |
| 129.11 | |
| Liquid |
| 174°C | |
| MFCD00007651 | |
| 2810 | |
| YNOOQIUSYGWMSS-UHFFFAOYSA-N | |
| 2,5-difluoroaniline | |
| 67775 | |
| ≥98.0% (GC) | |
| 2,5-Difluoroaniline |
Safety and Handling
TSCA : No