accessibility menu, dialog, popup

UserName

NH2FF

CAS RN 367-30-6

IUPAC Name: 2,5-difluoroaniline
Synonyms: 2,5-difluoroaniline
Molecular Weight (g/mol): 129.11
Molecular Formula: C6H5F2N
InChi Key: YNOOQIUSYGWMSS-UHFFFAOYSA-N
SMILES: NC1=CC(F)=CC=C1F
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)