missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluoroaniline, 98%

Catalog No. p-7080654
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1027209 10 g
AAA1027218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1027209 Supplier Thermo Scientific Chemicals Supplier No. A1027209

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 367-30-6 | C6H5F2N | 129.11 g/mol

2,5-Difluoroaniline was used in the synthesis of 2,4-di-tert-butyl-6-[(2,5-difluorophenyl)iminomethyl]phenol. It was also used in ultrasound-assisted preparation of 1,4-diazabutadienes. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Difluoroaniline was used in the synthesis of 2,4-di-tert-butyl-6-[(2,5-difluorophenyl)iminomethyl]phenol. It was also used in ultrasound-assisted preparation of 1,4-diazabutadienes. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Not miscible in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry condition in well sealed containers.

Chemical Identifiers

CAS 367-30-6
Molecular Formula C6H5F2N
Molecular Weight (g/mol) 129.11
MDL Number MFCD00007651
InChI Key YNOOQIUSYGWMSS-UHFFFAOYSA-N
Synonym benzenamine, 2,5-difluoro, 2,5-difluorobenzenamine, 2,5-difluoro-phenylamine, aniline, 2,5-difluoro, 2.5-difluoroaniline, aniline,5-difluoro, pubchem1531, 2,5-difluoro-aniline, 2,5-difluorophenylamine, benzenamine,5-difluoro
PubChem CID 67775
IUPAC Name 2,5-difluoroaniline
SMILES C1=CC(=C(C=C1F)N)F

Specifications

Melting Point 13°C to 14°C
Density 1.285
Boiling Point 171°C to 174°C
Flash Point 68°C (154°F)
Odor Characteristic
Refractive Index 1.513
Quantity 10 g
UN Number UN2941
Beilstein 2802549
Solubility Information Not miscible in water.
Formula Weight 129.11
Percent Purity 98%
Chemical Name or Material 2,5-Difluoroaniline
Show More Show Less
Hazard Category H227-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H311-H331-H373-H227-H302-H315-H319
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: FLUOROANILINES
EINECSNumber 206-690-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only