missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorophenol, 98%

Catalog No. AC113580500
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC113580500 50 g Glass bottle
AC113582500 250 g Glass bottle
AC113580010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC113580500 Supplier Thermo Scientific Chemicals Supplier No. 113580500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 583-78-8 | C6H4Cl2O | 163.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 583-78-8
Molecular Formula C6H4Cl2O
Molecular Weight (g/mol) 163.00
MDL Number MFCD00002174
InChI Key RANCECPPZPIPNO-UHFFFAOYSA-N
Synonym phenol, 2,5-dichloro, 2,5-dichloro-phenol, 2,5-dcp, unii-3b11g9akba, 3,6-dichlorophenol, ccris 5903, 3b11g9akba, dsstox_cid_5003, dsstox_rid_77621, dsstox_gsid_25003
PubChem CID 66
ChEBI CHEBI:27929
IUPAC Name 2,5-dichlorophenol
SMILES OC1=CC(Cl)=CC=C1Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 54.0°C to 58.0°C
Color White to Brown
Boiling Point 211.0°C
Flash Point 100°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula Cl2C6H3OH
Quantity 50 g
Beilstein 06,189
Solubility Information Solubility in water: 3g/L (20°C)
Formula Weight 163
Percent Purity 98%
Physical Form Crystals
Chemical Name or Material 2, 5-Dichlorophenol, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
EINECSNumber 209-520-4
RTECSNumber SK8600000
TSCA TSCA

RUO – Research Use Only