accessibility menu, dialog, popup

UserName

CAS RN 583-78-8

IUPAC Name: 2,5-dichlorophenol
Synonyms: 2,5-dichlorophenol
Molecular Weight (g/mol): 163
Molecular Formula: C6H4Cl2O
InChi Key: RANCECPPZPIPNO-UHFFFAOYSA-N
SMILES: OC1=CC(Cl)=CC=C1Cl
Boiling Point 
  • (4)
  • (3)
Percent Purity 
  • (1)
  • (6)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (6)
  • (4)
  • (3)
  • (1)
  • (6)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)