missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorohydroquinone, 97%

Catalog No. AAB2128503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2128503 1 g
AAB2128506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2128503 Supplier Thermo Scientific Chemicals Supplier No. B2128503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 824-69-1 | C6H4Cl2O2 | 179.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 824-69-1
Molecular Formula C6H4Cl2O2
Molecular Weight (g/mol) 179.00
MDL Number MFCD00041749
InChI Key AYNPIRVEWMUJDE-UHFFFAOYSA-N
Synonym 2,5-dichlorohydroquinone, 1,4-benzenediol, 2,5-dichloro, 2,5-dichloro-p-hydroquinone, hydroquinone, 2,5-dichloro, 2,5-dichloro-p-benzohydroquinone, 2,5-dchq, 2,5-dichloro-1,4-hydroquinone, 2,5-dichloro-1,4-benzenediol, unii-d489x4tqvd, 2,5-dichloro-1,4-dihydroxybenzene
PubChem CID 65
ChEBI CHEBI:27545
IUPAC Name 2,5-dichlorobenzene-1,4-diol
SMILES OC1=CC(Cl)=C(O)C=C1Cl

Specifications

Melting Point 168°C to 171°C
Quantity 1 g
Formula Weight 179
Percent Purity 97%
Chemical Name or Material 2,5-Dichlorohydroquinone
Hazard Category H314
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 212-533-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only