missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorobenzoyl Chloride 98.0+%, TCI America™

Catalog No. D29535G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29535G 5 g
D295325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D29535G Supplier TCI America Supplier No. D29535G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2905-61-5
Molecular Formula C7H3Cl3O
Molecular Weight (g/mol) 209.45
MDL Number MFCD00035821
InChI Key RSINFFVFOTUDEC-UHFFFAOYSA-N
PubChem CID 17945
IUPAC Name 2,5-dichlorobenzoyl chloride
SMILES ClC(=O)C1=CC(Cl)=CC=C1Cl

Specifications

Color White-Yellow
Boiling Point 95°C
Quantity 5 g
UN Number 3261
Formula Weight 209.45
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 2,5-Dichlorobenzoyl Chloride
EINECSNumber (3)-0062&(3)-1409
TSCA Yes