accessibility menu, dialog, popup

UserName

CAS RN 2905-61-5

IUPAC Name: 2,5-dichlorobenzoyl chloride
Synonyms: 2,5-dichlorobenzoyl chloride
Molecular Weight (g/mol): 209.45
Molecular Formula: C7H3Cl3O
InChi Key: RSINFFVFOTUDEC-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC(Cl)=CC=C1Cl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results