missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorobenzotrifluoride, 98%

Catalog No. p-7081365
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1131006 5 g
AAA1131014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1131006 Supplier Thermo Scientific Chemicals Supplier No. A1131006
Only null left

CAS: 320-50-3 | C7H3Cl2F3 | 214.996 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 320-50-3
Molecular Formula C7H3Cl2F3
Molecular Weight (g/mol) 214.996
MDL Number MFCD00000608
InChI Key DYBYUWVMLBBEMA-UHFFFAOYSA-N
Synonym 2,5-dichlorobenzotrifluoride, 1,4-dichloro-2-trifluoromethyl benzene, benzene, 1,4-dichloro-2-trifluoromethyl, 2,5-dichloro-alpha,alpha,alpha-trifluorotoluene, 2,5-dichloro-a,a,a-trifluorotoluene, toluene, 2,5-dichloro-alpha,alpha,alpha-trifluoro-6ci,7ci,8ci, pubchem3450, ksc925c7r, 2,5-dichloro trifluoromethylbenzene
PubChem CID 35360
IUPAC Name 1,4-dichloro-2-(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1Cl)C(F)(F)F)Cl

Specifications

Density 1.483
Flash Point 76°C (168°F)
Refractive Index 1.483
Quantity 5 g
UN Number UN1760
Beilstein 1955442
Formula Weight 215
Percent Purity 98%
Chemical Name or Material 2,5-Dichlorobenzotrifluoride
Hazard Category H227-H314
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
EINECSNumber 206-276-0
RTECSNumber CZ5553755
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only