missing translation for 'onlineSavingsMsg'
Learn More

2,5-dibromotoluene, Restek DFS Item

$117.30 - $169.28

Chemical Identifiers

CAS 615-59-8
Molecular Formula CH3OH
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Concentration Concentration or Composition Notes Price Quantity  
Catalog Number Mfr. No. Quantity Concentration Concentration or Composition Notes Price Quantity  
06728767 DFS Item
View Documents
Chromatographic
RK30435
1 mL 1000 μg/mL In P and T methanol N/A
Only null left
 
06728812 DFS Item
View Documents
Chromatographic
RK30453
1 mL 10,000 μg/mL In P and T methanol N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

615-59-8
CH3OH

Specifications

MA VPH Surrogate Standard
615-59-8
CH3OH
1 mL
Ampul
In P and T methanol
1000 μg/mL
99%
Videos
SDS
Documents

Documents