missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromoterephthalic Acid 98.0+%, TCI America™

Catalog No. p-7128037
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39945G 5 g
D399425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D39945G Supplier TCI America Supplier No. D39945G

Chemical Identifiers

CAS 13731-82-3
Molecular Formula C8H4Br2O4
Molecular Weight (g/mol) 323.92
MDL Number MFCD00204593
InChI Key VUTICWRXMKBOSF-UHFFFAOYSA-N
Synonym 2,5-dibromoterephtalic acid, 2,5-dibromobenzene-1,4-dicarboxylic acid, 1,4-benzenedicarboxylic acid, 2,5-dibromo, pubchem14888, 1, 2,5-dibromo, maybridge1_002153, 2,5-dibromoterephtalicacid, 2,5-dibromoterephthalicacid, acmc-1cb24, ksc174o0t
PubChem CID 83690
IUPAC Name 2,5-dibromobenzene-1,4-dicarboxylic acid
SMILES OC(=O)C1=CC(Br)=C(C=C1Br)C(O)=O

Specifications

Melting Point 318°C
Color White-Yellow
Quantity 5 g
Formula Weight 323.92
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dibromoterephthalic Acid
EINECSNumber (3)-1417&(3)-0062
TSCA Yes