accessibility menu, dialog, popup

UserName

CAS RN 13731-82-3

IUPAC Name: 2,5-dibromobenzene-1,4-dicarboxylic acid
Synonyms: 2,5-dibromobenzene-1,4-dicarboxylic acid
Molecular Weight (g/mol): 323.92
Molecular Formula: C8H4Br2O4
InChi Key: VUTICWRXMKBOSF-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(Br)=C(C=C1Br)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results