missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromonitrobenzene, 97%

Catalog No. AC112860250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC112860250 25 g Glass bottle
AC112861000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC112860250 Supplier Thermo Scientific Chemicals Supplier No. 112860250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3460-18-2 | C6H3Br2NO2 | 280.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3460-18-2
Molecular Formula C6H3Br2NO2
Molecular Weight (g/mol) 280.89
MDL Number MFCD00007046
InChI Key WRGKKASJBOREMB-UHFFFAOYSA-N
Synonym 2,5-dibromonitrobenzene, 1,4-dibromo-2-nitro-benzene, benzene, 1,4-dibromo-2-nitro, 2,5-dibromo-1-nitrobenzene, rarechem fh 1w 0042, pubchem3899, 1,4-dibromonitrobenzene, 2,5-dibromo nitrobenzene, 2,5-dibromo-nitrobenzene, acmc-1ahj1
PubChem CID 77004
IUPAC Name 1,4-dibromo-2-nitrobenzene
SMILES C1=CC(=C(C=C1Br)[N+](=O)[O-])Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 81°C to 85°C
Color Yellow
Density 2.3740g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula Br2C6H3NO2
Quantity 25 g
Beilstein 05, 250
Specific Gravity 2.374
Formula Weight 280.89
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2, 5-Dibromonitrobenzene, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 222-404-8
TSCA TSCA

RUO – Research Use Only