accessibility menu, dialog, popup

UserName

CAS RN 3460-18-2

IUPAC Name: 1,4-dibromo-2-nitrobenzene
Synonyms: 1,4-dibromo-2-nitro-benzene
Molecular Weight (g/mol): 280.9
Molecular Formula: C6H3Br2NO2
InChi Key: WRGKKASJBOREMB-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(Br)=CC=C1Br
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (2)
  • (3)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (4)