missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dibromohydroquinone 98.0+%, TCI America™
Supplier: TCI America D406725G
Specifications
| 2,5-Dibromohydroquinone | |
| 193°C | |
| MFCD00192664 | |
| 2,5-dibromohydroquinone, 2,5-dibromo-1,4-benzenediol, 2,5-dibromohydroquine, 1,4-benzenediol,2,5-dibromo, 2,5-dibromo-benzene-1,4-diol, 1,4-benzenediol, 2,5-dibromo, 2,5-dibromo-1,4-dihydroxybenzene | |
| C1=C(C(=CC(=C1Br)O)Br)O | |
| 267.904 | |
| 267.90 | |
| Crystalline Powder |
| 14753-51-6 | |
| C6H4Br2O2 | |
| 25 g | |
| VALXCIRMSIFPFN-UHFFFAOYSA-N | |
| 2,5-dibromobenzene-1,4-diol | |
| 280945 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 14753-51-6 | |
| 267.904 | |
| VALXCIRMSIFPFN-UHFFFAOYSA-N | |
| 2,5-dibromobenzene-1,4-diol |
| C6H4Br2O2 | |
| MFCD00192664 | |
| 280945 | |
| C1=C(C(=CC(=C1Br)O)Br)O |
Safety and Handling
TSCA : No