missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromohydroquinone 98.0+%, TCI America™

Catalog No. p-7128084
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40675G 5 g
D406725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D40675G Supplier TCI America Supplier No. D40675G
Only null left

Chemical Identifiers

CAS 14753-51-6
Molecular Formula C6H4Br2O2
Molecular Weight (g/mol) 267.904
MDL Number MFCD00192664
InChI Key VALXCIRMSIFPFN-UHFFFAOYSA-N
PubChem CID 280945
IUPAC Name 2,5-dibromobenzene-1,4-diol
SMILES C1=C(C(=CC(=C1Br)O)Br)O

Specifications

Melting Point 193°C
Quantity 5 g
Synonym 2,5-dibromohydroquinone, 2,5-dibromo-1,4-benzenediol, 2,5-dibromohydroquine, 1,4-benzenediol,2,5-dibromo, 2,5-dibromo-benzene-1,4-diol, 1,4-benzenediol, 2,5-dibromo, 2,5-dibromo-1,4-dihydroxybenzene
Formula Weight 267.90
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dibromohydroquinone