missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromobenzoic Acid 97.0+%, TCI America™

Catalog No. D34395G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D34395G 5 g
D343925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D34395G Supplier TCI America Supplier No. D34395G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 610-71-9
Molecular Formula C7H4Br2O2
Molecular Weight (g/mol) 279.92
MDL Number MFCD00016494
InChI Key SQQKOTVDGCJJKI-UHFFFAOYSA-N
Synonym benzoic acid, 2,5-dibromo, rarechem fh 1w 0049, pubchem3859, 2,5-dibromobenzoicacid, 2,5-dibromo-benzoic acid, benzoicacid, 2,5-dibromo, 4-09-00-01027 beilstein handbook reference, ksc353s4r, acmc-1b114, 2,5-dibromobenzoic acid
PubChem CID 11891
IUPAC Name 2,5-dibromobenzoic acid
SMILES OC(=O)C1=CC(Br)=CC=C1Br

Specifications

Melting Point 157°C
Quantity 5 g
Formula Weight 279.92
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dibromobenzoic Acid
EINECSNumber (3)-0062
RTECSNumber DG6290000
TSCA No