accessibility menu, dialog, popup

UserName

HOOBrBr

CAS RN 610-71-9

IUPAC Name: 2,5-dibromobenzoic acid
Synonyms: 2,5-dibromobenzoic acid
Molecular Weight (g/mol): 279.92
Molecular Formula: C7H4Br2O2
InChi Key: SQQKOTVDGCJJKI-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(Br)=CC=C1Br
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)