missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromobenzoic acid, 98%

Catalog No. p-7022352
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1507706 5 g
AAA1507714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1507706 Supplier Thermo Scientific Chemicals Supplier No. A1507706

CAS: 610-71-9 | C7H4Br2O2 | 279.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 610-71-9
Molecular Formula C7H4Br2O2
Molecular Weight (g/mol) 279.92
MDL Number MFCD00016494
InChI Key SQQKOTVDGCJJKI-UHFFFAOYSA-N
Synonym benzoic acid, 2,5-dibromo, rarechem fh 1w 0049, pubchem3859, 2,5-dibromobenzoicacid, 2,5-dibromo-benzoic acid, benzoicacid, 2,5-dibromo, 4-09-00-01027 beilstein handbook reference, ksc353s4r, acmc-1b114, 2,5-dibromobenzoic acid
PubChem CID 11891
IUPAC Name 2,5-dibromobenzoic acid
SMILES OC(=O)C1=CC(Br)=CC=C1Br

Specifications

Melting Point 153°C to 157°C
Quantity 5 g
Beilstein 1868193
Formula Weight 279.93
Percent Purity 98%
Chemical Name or Material 2,5-Dibromobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-234-7
RTECSNumber DG6290000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only