missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dibromoaniline 98.0+%, TCI America™
Supplier: TCI America D198925G
Specifications
| 2,5-Dibromoaniline | |
| 56°C | |
| MFCD00007636 | |
| 2,5-dibromobenzenamine, benzenamine, 2,5-dibromo, 2,5-dibromophenylamine, rarechem fh 1w 0043, pubchem3452, 2,5-dibromo-phenylamine, aniline, 2,5-dibromo, 2,5-bis bromanyl aniline, acmc-209im6, 2,5-dibromoaniline | |
| C1=CC(=C(C=C1Br)N)Br | |
| 250.921 | |
| 250.92 | |
| Crystalline Powder |
| 3638-73-1 | |
| C6H5Br2N | |
| 25 g | |
| WRTAZRGRFBCKBU-UHFFFAOYSA-N | |
| 2,5-dibromoaniline | |
| 77198 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 3638-73-1 | |
| 250.921 | |
| WRTAZRGRFBCKBU-UHFFFAOYSA-N | |
| 77198 | |
| C1=CC(=C(C=C1Br)N)Br |
| C6H5Br2N | |
| MFCD00007636 | |
| 2,5-dibromobenzenamine, benzenamine, 2,5-dibromo, 2,5-dibromophenylamine, rarechem fh 1w 0043, pubchem3452, 2,5-dibromo-phenylamine, aniline, 2,5-dibromo, 2,5-bis bromanyl aniline, acmc-209im6, 2,5-dibromoaniline | |
| 2,5-dibromoaniline |
Safety and Handling
TSCA : No