missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromoaniline 98.0+%, TCI America™

Catalog No. p-7127052
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
D19895G 5 g
D198925G 25 g
2 options

Catalog No. D19895G

Supplier: TCI America D19895G

Only null left

Chemical Identifiers

CAS 3638-73-1
Molecular Formula C6H5Br2N
Molecular Weight (g/mol) 250.921
MDL Number MFCD00007636
InChI Key WRTAZRGRFBCKBU-UHFFFAOYSA-N
Synonym 2,5-dibromobenzenamine, benzenamine, 2,5-dibromo, 2,5-dibromophenylamine, rarechem fh 1w 0043, pubchem3452, 2,5-dibromo-phenylamine, aniline, 2,5-dibromo, 2,5-bis bromanyl aniline, acmc-209im6, 2,5-dibromoaniline
PubChem CID 77198
IUPAC Name 2,5-dibromoaniline
SMILES C1=CC(=C(C=C1Br)N)Br

Specifications

Melting Point 56°C
Quantity 5 g
Formula Weight 250.92
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dibromoaniline