missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dibromo-p-xylene 98.0+%, TCI America™
Supplier: TCI America D1841100G
Specifications
| 2,5-Dibromo-p-xylene | |
| 1074-24-4 | |
| 150°C | |
| Crystal-Powder at 20°C | |
| MFCD00000091 | |
| QENIALCDPFDFHX-UHFFFAOYSA-N | |
| 1,4-dibromo-2,5-dimethylbenzene | |
| 66175 | |
| ≥98.0% (GC) |
| 75°C | |
| White-Yellow | |
| 100 g | |
| C8H8Br2 | |
| 2,5-dibromo-p-xylene, benzene, 1,4-dibromo-2,5-dimethyl, p-xylene, 2,5-dibromo, 2,5-dibromo-1,4-dimethylbenzene, 2,5-dimethyl-1,4-dibromobenzene, 2,5-dibromo-4-xylene, 2,4-dibromobenzene, p-xylene,5-dibromo, pubchem14829, acmc-1c2tg | |
| CC1=CC(Br)=C(C)C=C1Br | |
| 263.96 | |
| 263.96 |
Chemical Identifiers
| 1074-24-4 | |
| 263.96 | |
| QENIALCDPFDFHX-UHFFFAOYSA-N | |
| 66175 | |
| CC1=CC(Br)=C(C)C=C1Br |
| C8H8Br2 | |
| MFCD00000091 | |
| 2,5-dibromo-p-xylene, benzene, 1,4-dibromo-2,5-dimethyl, p-xylene, 2,5-dibromo, 2,5-dibromo-1,4-dimethylbenzene, 2,5-dimethyl-1,4-dibromobenzene, 2,5-dibromo-4-xylene, 2,4-dibromobenzene, p-xylene,5-dibromo, pubchem14829, acmc-1c2tg | |
| 1,4-dibromo-2,5-dimethylbenzene |
Safety and Handling
TSCA : Yes