missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromo-p-xylene 98.0+%, TCI America™

Catalog No. p-7126963
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D184125G 25 g
D1841100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D184125G Supplier TCI America Supplier No. D184125G
Only null left

Chemical Identifiers

CAS 1074-24-4
Molecular Formula C8H8Br2
Molecular Weight (g/mol) 263.96
MDL Number MFCD00000091
InChI Key QENIALCDPFDFHX-UHFFFAOYSA-N
Synonym 2,5-dibromo-p-xylene, benzene, 1,4-dibromo-2,5-dimethyl, p-xylene, 2,5-dibromo, 2,5-dibromo-1,4-dimethylbenzene, 2,5-dimethyl-1,4-dibromobenzene, 2,5-dibromo-4-xylene, 2,4-dibromobenzene, p-xylene,5-dibromo, pubchem14829, acmc-1c2tg
PubChem CID 66175
IUPAC Name 1,4-dibromo-2,5-dimethylbenzene
SMILES CC1=CC(Br)=C(C)C=C1Br

Specifications

Melting Point 75°C
Color White-Yellow
Boiling Point 150°C
Quantity 25 g
Physical Form Crystal-Powder at 20°C
Formula Weight 263.96
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2,5-Dibromo-p-xylene