missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dibromo-1,3-difluorobenzene 98.0+%, TCI America™
$78.29 - $240.46
Chemical Identifiers
| CAS | 128259-71-2 |
|---|---|
| Molecular Formula | C6H2Br2F2 |
| Molecular Weight (g/mol) | 271.887 |
| MDL Number | MFCD07783665 |
| InChI Key | OCUXAWVQPORIBP-UHFFFAOYSA-N |
| PubChem CID | 24721227 |
| IUPAC Name | 2,5-dibromo-1,3-difluorobenzene |
| SMILES | C1=C(C=C(C(=C1F)Br)F)Br |
Chemical Identifiers
| 128259-71-2 | |
| 271.887 | |
| OCUXAWVQPORIBP-UHFFFAOYSA-N | |
| 2,5-dibromo-1,3-difluorobenzene |
| C6H2Br2F2 | |
| MFCD07783665 | |
| 24721227 | |
| C1=C(C=C(C(=C1F)Br)F)Br |
Specifications
| 128259-71-2 | |
| C6H2Br2F2 | |
| 1 g | |
| C1=C(C=C(C(=C1F)Br)F)Br | |
| 271.887 | |
| 271.89 | |
| Crystalline Powder |
| 33°C | |
| MFCD07783665 | |
| OCUXAWVQPORIBP-UHFFFAOYSA-N | |
| 2,5-dibromo-1,3-difluorobenzene | |
| 24721227 | |
| ≥98.0% (GC) | |
| 2,5-Dibromo-1,3-difluorobenzene |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator