CAS RN 128259-71-2
CAS RN 128259-71-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2,5-Dibromo-1,3-difluorobenzene 98.0+%, TCI America™
CAS: 128259-71-2 Molecular Formula: C6H2Br2F2 Molecular Weight (g/mol): 271.887 MDL Number: MFCD07783665 InChI Key: OCUXAWVQPORIBP-UHFFFAOYSA-N PubChem CID: 24721227 IUPAC Name: 2,5-dibromo-1,3-difluorobenzene SMILES: C1=C(C=C(C(=C1F)Br)F)Br